Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Aminonicotinate 97.0+%, TCI America™
SDP

Catalog No. M21251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21251G 1 g
M21255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21251G Supplier TCI America Supplier No. M21251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14667-47-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00128872
InChI Key NZZDEODTCXHCRS-UHFFFAOYSA-N
PubChem CID 351633
IUPAC Name methyl 2-aminopyridine-3-carboxylate
SMILES COC(=O)C1=CC=CN=C1N

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 1 g
Synonym methyl 2-aminonicotinate, 2-amino-nicotinic acid methyl ester, 2-aminonicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl2-aminonicotinate, pubchem16778, methyl?2-aminonicotinate, methyl 2-amino-nicotinate, acmc-1c0pk, aminonicotinsauremethylester
Formula Weight 152.15
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Aminonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.