Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-aminopyridine-4-carboxylate, 97%

Catalog No. AAL1986903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1986903 1 g
AAL1986906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1986903 Supplier Thermo Scientific Chemicals Supplier No. L1986903
Add to Cart

CAS: 6937-03-7 | C7H8N2O2 | 152.15 g/mol

Methyl 2-aminopyridine-4-carboxylate is an inhibitor of nitrogen monoxide synthetase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-aminopyridine-4-carboxylate is an inhibitor of nitrogen monoxide synthetase.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6937-03-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD04039316
InChI Key SVWWNEYBEFASMP-UHFFFAOYSA-N
Synonym methyl 2-aminoisonicotinate, 2-aminopyridine-4-carboxylic acid methyl ester, 2-aminoisonicotinic acid methyl ester, 4-methoxycarbonyl pyridin-2-amine, 4-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl 2-amino-4-pyridinecarboxylate, methyl-2-aminopyridine-4-carboxylate, 2-amino-4-pyridinecarboxylic acid methylester, 2-amino-4-pyridinecarboxylic acid methyl ester, methyl-2-aminoisonicotinat
PubChem CID 236984
IUPAC Name methyl 2-aminopyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC=C1)N

Specifications

Melting Point 145°C to 149°C
Quantity 1 g
Beilstein 128642
Solubility Information Insoluble in water.
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material Methyl 2-aminopyridine-4-carboxylate
Hazard Category H302+H332-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.