Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 2-bromo-2-butenoate

Catalog No. 165035ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-503-5ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-503-5ML Supplier Sigma Aldrich Supplier No. 165035ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl 2-bromo-2-butenoate

Specifications

CAS 17642-18-1
Density 1.505 g/mL (at 20°C (literature))
Boiling Point 178°C to 182°C (lit.)
Molecular Formula C5H7BrO2
Refractive Index n20/D 1.486
Linear Formula CH3CH=CBrCOOCH3
MDL Number MFCD00042891
Quantity 1 g
Synonym Methyl 2-bromocrotonate
Molecular Weight (g/mol) 179.01
Percent Purity ≥95.0% (GC)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.