Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-bromo-6-fluorophenylacetate, 96%, Thermo Scientific™

Catalog No. AAH6246503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6246503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6246503 Supplier Thermo Scientific Chemicals Supplier No. H6246503

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1427397-15-6
Molecular Formula C9H8BrFO2
Molecular Weight (g/mol) 247.063
InChI Key QXIUUACWTNNJID-UHFFFAOYSA-N
Synonym methyl 2-2-bromo-6-fluorophenyl acetate
PubChem CID 100912176
IUPAC Name methyl 2-(2-bromo-6-fluorophenyl)acetate
SMILES COC(=O)CC1=C(C=CC=C1Br)F

Specifications

Quantity 1 g
Formula Weight 247.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 2-bromo-6-fluorophenylacetate
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.