Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-bromobenzoate, 98%, Thermo Scientific™

Catalog No. AC249811000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC249811000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC249811000 Supplier Thermo Scientific Chemicals Supplier No. 249811000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-94-6
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00016328
InChI Key SWGQITQOBPXVRC-UHFFFAOYSA-N
Synonym methyl o-bromobenzoate, 2-bromobenzoic acid methyl ester, benzoic acid, 2-bromo-, methyl ester, methyl-2-bromobenzoate, methyl bromobenzoate, o-bromobenzoic acid, methyl ester, benzoic acid, o-bromo-, methyl ester, pubchem3956, 2-bromo methyl benzoate, acmc-1atow
PubChem CID 11894
IUPAC Name methyl 2-bromobenzoate
SMILES COC(=O)C1=CC=CC=C1Br

Specifications

Density 1.5100g/mL
Boiling Point 252.0°C to 254.0°C
Refractive Index 1.5575 to 1.5595
Linear Formula BrC6H4CO2CH3
Quantity 100 mL
Specific Gravity 1.51
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in polar solvents, insoluble in non-polar solvents
Formula Weight 215.05
Chemical Name or Material Methyl 2-bromobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.