Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 2-(bromomethyl)acrylate | 4224-69-5 | MFCD00011697 | 99.4% | 179.01 g/mol | C5H7BrO2 | 25 G
SDP

Catalog No. 5000303097
Encompass_Preferred

Methyl 2-(bromomethyl)acrylate is an organic reagent used in biochemical and medicinal chemistry research as a reactive building block and assay reagent. It is a methyl ester bearing a bromomethyl-substituted acrylate group and is supplied with formulation and stability data for research applications.

  • High purity (≈99.4%).
  • Molecular formula C5H7BrO2; molecular weight 179.01 g/mol.
  • Readily soluble in DMSO (200 mg/mL; ultrasonic assistance may be required).
  • Available as solid and as prepared DMSO solutions for convenience in assays.
  • Formulatable for in vivo dosing at ≥5 mg/mL in common vehicle mixtures.
  • Store at 4°C, protect from light; solutions stable at -80°C for extended storage.

Catalog No. 50-003-03097 Supplier Medchemexpress LLC Supplier No. 25GHYW016582
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 2-(bromomethyl)acrylate is an organic reagent used in biochemical and medicinal chemistry research as a reactive building block and assay reagent. It is a methyl ester bearing a bromomethyl-substituted acrylate group and is supplied with formulation and stability data for research applications.

  • High purity (≈99.4%).
  • Molecular formula C5H7BrO2; molecular weight 179.01 g/mol.
  • Readily soluble in DMSO (200 mg/mL; ultrasonic assistance may be required).
  • Available as solid and as prepared DMSO solutions for convenience in assays.
  • Formulatable for in vivo dosing at ≥5 mg/mL in common vehicle mixtures.
  • Store at 4°C, protect from light; solutions stable at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.