Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Bromonicotinate 98.0+%, TCI America™
SDP

Catalog No. M24491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24491G 1 g
M24495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24491G Supplier TCI America Supplier No. M24491G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52718-95-3
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD07367915
InChI Key RAFFKXWNTXTTFO-UHFFFAOYSA-N
Synonym 2-Bromonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-3-carboxylate, 2-Bromopyridine-3-carboxylic Acid Methyl Ester
PubChem CID 12349191
IUPAC Name methyl 2-bromopyridine-3-carboxylate
SMILES COC(=O)C1=CC=CN=C1Br

Specifications

Quantity 1 g
Formula Weight 216.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 2-Bromonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.