Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 2-bromopropionate

Catalog No. 167185100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-718-5100G 100 g
16-718-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-718-5100G Supplier Sigma Aldrich Supplier No. 167185100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl 2-bromopropionate was used as initiator during the synthesis of polymer brushes with single-walled carbon nanotubes as backbones
  • It was used as starting reagent for the synthesis of 2-18f-fluoropropionic acid, positron emission tomography imaging agent for prostate cancer

Specifications

CAS 5445-17-0
Density 1.497 g/mL (at 25°C (literature))
Boiling Point 51°C (19 mmHg)
Molecular Formula C4H7BrO2
Refractive Index n20/D 1.451 (literature)
Linear Formula CH3CHBrCO2CH3
Quantity 100 g
MDL Number MFCD00000143
Molecular Weight (g/mol) 167
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.