Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Bromothiazole-5-carboxylate 95.0+%, TCI America™
SDP

Catalog No. M26695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26695G 5 g
M26691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26695G Supplier TCI America Supplier No. M26695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54045-74-8
Molecular Formula C5H4BrNO2S
Molecular Weight (g/mol) 222.056
MDL Number MFCD03788565
InChI Key HNLVIKMXFBRZDF-UHFFFAOYSA-N
Synonym methyl 2-bromothiazole-5-carboxylate, methyl 2-bromo-5-thiazole carboxylate, methyl2-bromothiazole-5-carboxylate, 5-thiazolecarboxylic acid, 2-bromo-, methyl ester, 2-bromo-thiazole-5-carboxylic acid methyl ester, 2-bromothiazole-5-carboxylic acid methyl ester, 2-bromo-1,3-thiazole-5-carboxylic acid methyl ester, pubchem17197, acmc-1apjm, ksc494s7p
PubChem CID 2773502
IUPAC Name methyl 2-bromo-1,3-thiazole-5-carboxylate
SMILES COC(=O)C1=CN=C(S1)Br

Specifications

Melting Point 62°C
Quantity 5 g
Formula Weight 222.06
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Bromothiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.