Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-chloro-4-fluorophenylacetate, 98%

Catalog No. AC304780050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC304780050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC304780050 Supplier Thermo Scientific Chemicals Supplier No. 304780050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 214262-88-1 | C9H8ClFO2 | 202.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214262-88-1
Molecular Formula C9H8ClFO2
Molecular Weight (g/mol) 202.61
MDL Number MFCD00800611
InChI Key DFTYYBRLWCAUHP-UHFFFAOYSA-N
Synonym methyl 2-2-chloro-4-fluorophenyl acetate, methyl 2-chloro-4-fluorophenylacetate, benzeneacetic acid, 2-chloro-4-fluoro-, methyl ester, methyl-2-chloro-4-fluorophenylacetate, pubchem16488, methyl 2-chloro-4-fluorophenyl acetate, methyl2-2-chloro-4-fluorophenyl acetate, 2-chloro-4-fluorophenyl-acetic acid methyl ester, 2-chloro-4-fluorophenyl acetic acid methyl ester, methyl 2-chloro-4-fluorophenylacetate 98
PubChem CID 736123
IUPAC Name methyl 2-(2-chloro-4-fluorophenyl)acetate
SMILES COC(=O)CC1=CC=C(F)C=C1Cl

Specifications

Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.5060 to 1.5080 (20°C, 589nm)
Linear Formula ClC6H3(F)CHCOCH3
Quantity 5 g
Formula Weight 202.51
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 2-chloro-4-fluorophenylacetate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.