Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Chloro-6-methylisonicotinate 97.0+%, TCI America™
SDP

Catalog No. M22395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22395G 5 g
M22391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22395G Supplier TCI America Supplier No. M22395G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3998-90-1
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD04038391
InChI Key BDWMGYZSQKGUFA-UHFFFAOYSA-N
Synonym methyl 2-chloro-6-methylisonicotinate, methyl 2-chloro-6-picoline-4-carboxylate, 2-chloro-6-picoline-4-carboxylic acid methyl ester, 2-chloro-6-methylpyridine-4-carboxylic acid methyl ester, 2-chloro-4-methoxycarbonyl-6-methylpyridine, methyl 6-chloro-2-methylpyridine-4-carboxylate, 2-chloro-6-methylisonicotinic acid methyl ester, 2-chloro-6-methyl pyridine-4-carboxylic acid methyl ester, 4-pyridinecarboxylic acid, 2-chloro-6-methyl-, methyl ester, pubchem17060
PubChem CID 2759833
IUPAC Name methyl 2-chloro-6-methylpyridine-4-carboxylate
SMILES COC(=O)C1=CC(C)=NC(Cl)=C1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 125°C
Quantity 5 g
Formula Weight 185.61
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Chloro-6-methylisonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.