Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Chloro-L-mandelate 98.0+%, TCI America™
SDP

Catalog No. M23831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M23831G Supplier TCI America Supplier No. M23831G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32345-60-1
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
InChI Key ZMPGBVQQIQSQED-QMMMGPOBSA-N
PubChem CID 11183258
IUPAC Name methyl (2S)-2-(2-chlorophenyl)-2-hydroxyacetate
SMILES COC(=O)C(C1=CC=CC=C1Cl)O

Specifications

Color Yellow
Quantity 1 g
Physical Form Clear Liquid at 20°C
Synonym 2-Chloro-L-mandelic Acid Methyl Ester, Methyl (S)-2′C-Chloro-alpha-hydroxyphenylacetate, (S)-2′C-Chloro-alpha-hydroxyphenylacetic Acid Methyl Ester
Formula Weight 200.62
Percent Purity ≥98.0% (GC)
Chemical Name or Material Methyl 2-Chloro-L-mandelate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.