Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 2-chlorooxazole-4-carboxylate

Catalog No. SYX003781G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003781G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003781G Supplier Sigma Aldrich Supplier No. SYX003781G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-chlorobenzyl bromide reacts with aminoethanol and nah to yield aminoethyl 3-chlorobenzyl ether.3-chlorobenzyl bromide was used in the synthesis of symmetrical and unsymmetrical benzyl thioethers
  • It was used as starting reagent during the synthesis of 1-(3-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1h-benzimidazole dihydrochloride

Specifications

CAS 934236-35-8
Molecular Formula C5H4ClNO3
Linear Formula C5H4ClNO3
MDL Number MFCD09265505
Quantity 10 g
Molecular Weight (g/mol) 161.54
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.