Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-cyanoisonicotinate, 97%, Thermo Scientific™

Catalog No. AAH6309003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6309003 1 g
AAH6309006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6309003 Supplier Thermo Scientific Chemicals Supplier No. H6309003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 2-cyanoisonicotinate can be used in the synthesis of various pharmaceutical and biologically active compounds, such as tetra substituted imidazolines as potent and selective neuropeptide Y (NPY) Y5 receptor antagonists.

Solubility
Slightly Soluble in water (6.2 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94413-64-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD07367894
InChI Key ORVHMLCJEKDDAX-UHFFFAOYSA-N
Synonym methyl 2-cyanoisonicotinate, 2-cyano-4-pyridine carboxylic acid methyl ester, 4-pyridinecarboxylicacid, 2-cyano-, methyl ester, 4-pyridinecarboxylic acid, 2-cyano-, methyl ester, methyl 2-cyanois, methyl 2-cyanoisonicotite, 2-cyano-4-carbomethoxypyridine, 2-cyanoisonicotinic acid methyl ester, 2-cyano-isonicotinic acid methyl ester, 2-cyano-4-pyridine-carboxylic acid methyl ester
PubChem CID 12132796
IUPAC Name methyl 2-cyanopyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC=C1)C#N

Specifications

Color Brown
Quantity 1 g
UN Number UN3439
Solubility Information Slightly Soluble in water (6.2g/L) (25°C).
Formula Weight 162.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 2-cyanoisonicotinate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.