Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 2-(diethoxyphosphoryl)acetate | 1067-74-9 | MFCD00009081 | 99.1% | 210.17 g/mol | C7H15O5P | 50 G
SDP

Catalog No. 5000300643
Encompass_Preferred

Methyl 2-(diethoxyphosphoryl)acetate (methyl diethylphosphonoacetate) is an organophosphorus biochemical reagent used in organic synthesis and life-science research. It is employed as a reactant for preparing branched allylic fluorides and in regioselective Diels-Alder reactions to access antibiotic building blocks and related intermediates. Supplied with characterization and safety documentation for laboratory research use only.

  • High purity (≈99.1% by supplier COA).
  • Molecular formula C7H15O5P; molecular weight 210.17 g/mol.
  • Supplied in laboratory-scale packages (50 g and 100 g).
  • Includes COA and SDS for batch verification and safe handling.
  • Recommended storage: pure form -20 °C (3 years) or 4 °C (2 years).
  • Suitable for reagent-grade synthetic applications in vitro.

Catalog No. 50-003-00643 Supplier Medchemexpress LLC Supplier No. HYW01845250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 2-(diethoxyphosphoryl)acetate (methyl diethylphosphonoacetate) is an organophosphorus biochemical reagent used in organic synthesis and life-science research. It is employed as a reactant for preparing branched allylic fluorides and in regioselective Diels-Alder reactions to access antibiotic building blocks and related intermediates. Supplied with characterization and safety documentation for laboratory research use only.

  • High purity (≈99.1% by supplier COA).
  • Molecular formula C7H15O5P; molecular weight 210.17 g/mol.
  • Supplied in laboratory-scale packages (50 g and 100 g).
  • Includes COA and SDS for batch verification and safe handling.
  • Recommended storage: pure form -20 °C (3 years) or 4 °C (2 years).
  • Suitable for reagent-grade synthetic applications in vitro.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.