Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Methyl 2-(dodecylthiocarbonothioylthio)-2-methylpropionate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene; acrylate and acrylamide monomers. Chain Transfrer Agent (CTA)
Specifications
Specifications
| CAS | 1088555-95-6 |
| Density | 1.008 g/mL (at 25°C) |
| Molecular Formula | C18H34O2S3 |
| Refractive Index | n20/D 1.517 |
| Linear Formula | C18H34O2S3 |
| Quantity | 5 g |
| MDL Number | MFCD19687018 |
| Synonym | 2-[[(Dodecylthio)thioxomethyl]thio]-2-methylpropanoic acid methyl ester |
| Molecular Weight (g/mol) | 378.66 |
| Percent Purity | 97% (HPLC) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?