Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Ethoxybenzoate 99.0+%, TCI America™
SDP

Catalog No. M178525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M178525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M178525G Supplier TCI America Supplier No. M178525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3686-55-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00017261
InChI Key QAYQKAPOTVSWLS-UHFFFAOYSA-N
Synonym 2-Ethoxybenzoic Acid Methyl Ester
PubChem CID 347794
IUPAC Name methyl 2-ethoxybenzoate
SMILES CCOC1=CC=CC=C1C(=O)OC

Specifications

Color Yellow
Boiling Point 245°C
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Ethoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.