Learn More
Methyl 2-fluorophenylacetate, 99%, Thermo Scientific™
Specifications
Specifications
| Color | Colorless |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4890 to 1.4910 |
| Quantity | 5 g |
| Formula Weight | 168.06 |
| Percent Purity | 99% |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl 2-fluorophenylacetate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.