Learn More
Medchemexpress LLC Methyl 2-furoate | 611-13-2 | MFCD00003236 | 99.9% | 126.11 g/mol | C6H6O3 | 500g
SDP
Catalog No. 5000267787
Encompass_Preferred
Methyl 2-furoate (Methyl furan-2-carboxylate) can be used to synthesize cis-fused 5-oxofuro[2 3-b]furans Methyl 2-furoate is also used in the preparation of sesquiterpene lactones[1]
Catalog No. 50-002-67787 Supplier Medchemexpress LLC Supplier No. HYY0949500G
Methyl 2-furoate (Methyl furan-2-carboxylate) can be used to synthesize cis-fused 5-oxofuro[2 3-b]furans Methyl 2-furoate is also used in the preparation of sesquiterpene lactones[1]