Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Hydroxy-2-methoxyacetate (contains related compounds), TCI America™
SDP

Catalog No. H070625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H070625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H070625G Supplier TCI America Supplier No. H070625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19757-97-2
Molecular Formula C4H8O4
Molecular Weight (g/mol) 120.104
MDL Number MFCD00043941
InChI Key OVJJVYHDJVQFSF-UHFFFAOYSA-N
Synonym Glyoxylic Acid Methyl Ester Methyl Hemiacetal, 2-Hydroxy-2-methoxyacetic Acid Methyl Ester
PubChem CID 2724791
IUPAC Name methyl 2-hydroxy-2-methoxyacetate
SMILES COC(C(=O)OC)O

Specifications

Color Colorless
Quantity 25 g
Formula Weight 120.10
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Methyl 2-Hydroxy-2-methoxyacetate (contains related compounds)
EINECSNumber (2)-3681
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.