Learn More
Matrix Scientific Methyl 2-iodobenzoate, 610-97-9, MFCD00016351, 100g
Supplier: Matrix Scientific 004193100G
Molecular Formula C8H7IO2, Purity 97%, Molecular Weight 262.05, Boiling Point 149-150/10mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.