Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-methylnicotinate, 97%

Catalog No. AAL0907406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0907406 5 g
AAL0907414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0907406 Supplier Thermo Scientific Chemicals Supplier No. L0907406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 65719-09-7 | C8H9NO2 | 151.165 g/mol

Methyl 2-Methylnicotinate is an ester derivative of nicotinic acid used in the preparation of bicyclic nitrogen containing heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-Methylnicotinate is an ester derivative of nicotinic acid used in the preparation of bicyclic nitrogen containing heterocycles.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65719-09-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00051961
InChI Key KLHWBYHFWALOIJ-UHFFFAOYSA-N
PubChem CID 7015327
IUPAC Name methyl 2-methylpyridine-3-carboxylate
SMILES CC1=C(C=CC=N1)C(=O)OC

Specifications

Boiling Point 102°C to 104°C (10 mmHg)
Refractive Index 1.5165
Quantity 5 g
Beilstein 471919
Synonym methyl 2-methylnicotinate, 2-methylnicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2-methyl-, methyl ester, methyl2-methylnicotinate, pubchem2218, acmc-1belg, methyi 2-methyinicotinate, methyl 2-methyinicotinate, methyl-2-methylnicotinate, ksc493g0t
Solubility Information Soluble in chloroform and methanol.
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material Methyl 2-methylnicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.