Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-methylnicotinate, 97%

Catalog No. AC449250050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449250050 5 g Glass bottle
AC449250250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449250050 Supplier Thermo Scientific Chemicals Supplier No. 449250050
Add to Cart

CAS: 65719-09-7 | C8H9NO2 | 151.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65719-09-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
InChI Key KLHWBYHFWALOIJ-UHFFFAOYSA-N
PubChem CID 7015327
IUPAC Name methyl 2-methylpyridine-3-carboxylate
SMILES CC1=C(C=CC=N1)C(=O)OC

Specifications

Boiling Point 102.0°C to 104.0°C (10.0 mmHg)
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym methyl 2-methylnicotinate, 2-methylnicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2-methyl-, methyl ester, methyl2-methylnicotinate, pubchem2218, acmc-1belg, methyi 2-methyinicotinate, methyl 2-methyinicotinate, methyl-2-methylnicotinate, ksc493g0t
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material Methyl 2-methylnicotinate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.