Learn More
Medchemexpress LLC Methyl 2-Octynoate | 111-12-6 | 99.8% | 154.21 | 25 G
SDP
Catalog No. 5000454011
Encompass_Preferred
Methyl 2-Octynoate is a hapten and a laboratory chemical used in the manufacture of substances. It is intended for research use only.
  • Acts as a hapten.
  • Used in laboratory research.
  • Aids in the manufacture of various substances.
  • Is a combustible liquid.
  • May cause an allergic skin reaction.
  • Very toxic to aquatic life.
  • Harmful to aquatic life with long-lasting effects.
  • Stable under recommended storage conditions.
  • Appearance is a colorless to light yellow liquid.
Catalog No. 50-004-54011 Supplier Medchemexpress LLC Supplier No. 25GHYW013576
Methyl 2-Octynoate is a hapten and a laboratory chemical used in the manufacture of substances. It is intended for research use only.
  • Acts as a hapten.
  • Used in laboratory research.
  • Aids in the manufacture of various substances.
  • Is a combustible liquid.
  • May cause an allergic skin reaction.
  • Very toxic to aquatic life.
  • Harmful to aquatic life with long-lasting effects.
  • Stable under recommended storage conditions.
  • Appearance is a colorless to light yellow liquid.