Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-octynoate, 98%

Catalog No. AAB2124214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2124214 25 g
AAB2124222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2124214 Supplier Thermo Scientific Chemicals Supplier No. B2124214
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 111-12-6 | C9H14O2 | 154.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-12-6
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD00009530
InChI Key FRLZQXRXIKQFNS-UHFFFAOYSA-N
Synonym methyl 2-octynoate, folione, methyl heptine carbonate, methyl 2-octynate, methyl 2-octinate, 2-octynoic acid, methyl ester, vert de violette, artificial, methyl hept-1-yne-1-carboxylate, methyl pentylacetylenecarboxylate, unii-0ttp6yt2t3
PubChem CID 8092
IUPAC Name methyl oct-2-ynoate
SMILES CCCCCC#CC(=O)OC

Specifications

Density 0.92
Boiling Point 216°C to 218°C
Flash Point 88°C (192°F)
Refractive Index 1.446
Quantity 25 g
Beilstein 1756887
Formula Weight 154.2
Percent Purity 98%
Chemical Name or Material Methyl 2-octynoate
Hazard Category H227-H317
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c
EINECSNumber 203-836-6
RTECSNumber RI2735000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.