Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Oxoindoline-6-carboxylate 98.0+%, TCI America™
SDP

Catalog No. M27991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27991G 1 g
M27995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27991G Supplier TCI America Supplier No. M27991G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14192-26-8
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD03095196
InChI Key YFTGUNWFFVDLNM-UHFFFAOYSA-N
Synonym 2-Oxoindoline-6-carboxylic Acid Methyl Ester
PubChem CID 3734372
IUPAC Name methyl 2-oxo-1,3-dihydroindole-6-carboxylate
SMILES COC(=O)C1=CC2=C(CC(=O)N2)C=C1

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 1 g
Formula Weight 191.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Oxoindoline-6-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.