Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,3,3-trifluoropyruvate, 97%

Catalog No. AC379710050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC379710050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC379710050 Supplier Thermo Scientific Chemicals Supplier No. 379710050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13089-11-7 | C4H3F3O3 | 156.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13089-11-7
Molecular Formula C4H3F3O3
Molecular Weight (g/mol) 156.06
MDL Number MFCD00114936
InChI Key XGLLQDIWQRQROJ-UHFFFAOYSA-N
Synonym methyl trifluoropyruvate, methyl 3,3,3-trifluoropyruvate, methyltrifluoropyruvate, trifluoropyruvic acid methyl ester, 3,3,3-trifluoropyruvic acid methyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, methyl ester, pubchem2012, rarechem al bf 1300, acmc-1bp0j
PubChem CID 2734931
IUPAC Name methyl 3,3,3-trifluoro-2-oxopropanoate
SMILES COC(=O)C(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.3900g/mL
Boiling Point 84°C to 86°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3320 to 1.334
Linear Formula F3CC(O)CO2CH3
Quantity 5 g
Specific Gravity 1.39
Formula Weight 156.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 3, 3, 3-trifluoropyruvate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.