Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 3,3-dimethoxypropionate | 7424-91-1 | MFCD00010650 | 100.0% | 148.16 g/mol | C6H12O4 | 25 G
SDP

Catalog No. 5000302713
Encompass_Preferred

Methyl 3,3-dimethoxypropionate is an organic ester used as a biochemical reagent and a synthetic building block in laboratory research, commonly employed in organic synthesis and life-science studies. It is typically supplied as a high-purity material for analytical and preparative applications.

  • High purity suitable for synthesis and analytical applications.
  • Versatile building block for organic synthesis.
  • Compatible with common laboratory handling and storage conditions.
  • Available in small pack sizes for benchtop work and scale-up.
  • Molecular formula C6H12O4; molecular weight 148.16 g/mol.
  • CAS number 7424-91-1 for substance identification.

Catalog No. 50-003-02713 Supplier Medchemexpress LLC Supplier No. 25GHYW017474
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 3,3-dimethoxypropionate is an organic ester used as a biochemical reagent and a synthetic building block in laboratory research, commonly employed in organic synthesis and life-science studies. It is typically supplied as a high-purity material for analytical and preparative applications.

  • High purity suitable for synthesis and analytical applications.
  • Versatile building block for organic synthesis.
  • Compatible with common laboratory handling and storage conditions.
  • Available in small pack sizes for benchtop work and scale-up.
  • Molecular formula C6H12O4; molecular weight 148.16 g/mol.
  • CAS number 7424-91-1 for substance identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.