Learn More
Medchemexpress LLC Methyl 3,3-dimethoxypropionate | 7424-91-1 | MFCD00010650 | ≥98.0% | 148.16 g·mol^-1 | C6H12O4 | 100 G
SDP
Catalog No. 5000300375
Encompass_Preferred
Methyl 3,3-dimethoxypropionate is an organic methyl ester used as a building block and intermediate in organic synthesis and life-science research. It is commonly employed in the preparation of derivatives and in multi-step synthetic sequences where a protected aldehyde equivalent is required.
  • High purity (≥98.0%) for research and synthesis.
  • Serves as a methyl ester building block in organic synthesis.
  • Useful as a protected aldehyde equivalent in multi-step syntheses.
  • Available in multiple pack sizes for lab-scale work.
  • Molecular formula C6H12O4 and molecular weight 148.16 g·mol^-1.
Catalog No. 50-003-00375 Supplier Medchemexpress LLC Supplier No. HYW017474100G
Methyl 3,3-dimethoxypropionate is an organic methyl ester used as a building block and intermediate in organic synthesis and life-science research. It is commonly employed in the preparation of derivatives and in multi-step synthetic sequences where a protected aldehyde equivalent is required.
  • High purity (≥98.0%) for research and synthesis.
  • Serves as a methyl ester building block in organic synthesis.
  • Useful as a protected aldehyde equivalent in multi-step syntheses.
  • Available in multiple pack sizes for lab-scale work.
  • Molecular formula C6H12O4 and molecular weight 148.16 g·mol^-1.