Learn More
Medchemexpress LLC Methyl 3,3-dimethoxypropionate | 7424-91-1 | MFCD00010650 | ≥98.0% | 148.16 g/mol | C6H12O4 | 250 G
SDP
Catalog No. 5000303616
Encompass_Preferred
Methyl 3,3-dimethoxypropionate is an organic methyl ester used as a reagent and synthetic intermediate in organic and medicinal chemistry. The molecule features a dimethoxy (acetal)-protected propionate unit that acts as a masked aldehyde equivalent, enabling controlled deprotection and further functionalization. It is supplied as a colorless liquid at research-grade purity for laboratory synthesis and method development.
  • Functions as a masked aldehyde equivalent in synthesis.
  • Provides an acetal-protected propionate for selective transformations.
  • Facilitates ester hydrolysis and downstream derivatization.
  • Suitable for multistep organic and medicinal chemistry workflows.
  • Available in research-scale quantities with high purity.
Catalog No. 50-003-03616 Supplier Medchemexpress LLC Supplier No. 250GHYW017474
Methyl 3,3-dimethoxypropionate is an organic methyl ester used as a reagent and synthetic intermediate in organic and medicinal chemistry. The molecule features a dimethoxy (acetal)-protected propionate unit that acts as a masked aldehyde equivalent, enabling controlled deprotection and further functionalization. It is supplied as a colorless liquid at research-grade purity for laboratory synthesis and method development.
  • Functions as a masked aldehyde equivalent in synthesis.
  • Provides an acetal-protected propionate for selective transformations.
  • Facilitates ester hydrolysis and downstream derivatization.
  • Suitable for multistep organic and medicinal chemistry workflows.
  • Available in research-scale quantities with high purity.