Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 3,3-dimethoxypropionate | 7424-91-1 | MFCD00010650 | ≥98.0% | 148.16 g/mol | C6H12O4 | 50 G
SDP

Catalog No. 5000299981
Encompass_Preferred

Methyl 3,3-dimethoxypropionate is the methyl ester of 3,3-dimethoxypropionic acid, provided as a high-purity biochemical reagent for organic synthesis and life-science research. It functions as a synthetic building block and intermediate for derivatization and coupling reactions.

  • High purity ≥98.0%.
  • Molecular formula C6H12O4.
  • Molecular weight 148.16 g/mol.
  • Available in multiple pack sizes including 50 g.
  • Suitable as a building block for organic synthesis and derivatization.

Catalog No. 50-002-99981 Supplier Medchemexpress LLC Supplier No. HYW01747450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 3,3-dimethoxypropionate is the methyl ester of 3,3-dimethoxypropionic acid, provided as a high-purity biochemical reagent for organic synthesis and life-science research. It functions as a synthetic building block and intermediate for derivatization and coupling reactions.

  • High purity ≥98.0%.
  • Molecular formula C6H12O4.
  • Molecular weight 148.16 g/mol.
  • Available in multiple pack sizes including 50 g.
  • Suitable as a building block for organic synthesis and derivatization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.