Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 3,3-dimethoxypropionate | 7424-91-1 | MFCD00010650 | >98.0% | 148.16 g/mol | C6H12O4 | 500 G
SDP

Catalog No. 5000301416
Encompass_Preferred

Methyl 3,3-dimethoxypropionate is an organic methyl ester used as a reagent and building block in organic synthesis and life-science research. It is provided as a high-purity material with documented physical data and recommended storage conditions to preserve stability.

  • Used as a synthetic building block in organic chemistry.
  • High purity (>98%) supports analytical and preparative work.
  • Molecular weight 148.16 g/mol and formula C6H12O4 for stoichiometric calculations.
  • Soluble in DMSO (10 mM) for solution-phase assays.
  • Recommended storage at -20°C (long term) or 4°C (short term) as powder.
  • Intended for research use only.

Catalog No. 50-003-01416 Supplier Medchemexpress LLC Supplier No. HYW017474500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 3,3-dimethoxypropionate is an organic methyl ester used as a reagent and building block in organic synthesis and life-science research. It is provided as a high-purity material with documented physical data and recommended storage conditions to preserve stability.

  • Used as a synthetic building block in organic chemistry.
  • High purity (>98%) supports analytical and preparative work.
  • Molecular weight 148.16 g/mol and formula C6H12O4 for stoichiometric calculations.
  • Soluble in DMSO (10 mM) for solution-phase assays.
  • Recommended storage at -20°C (long term) or 4°C (short term) as powder.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.