Learn More
Medchemexpress LLC HY-Z0548 50mg Medchemexpress, Methyl 3,4-dihydroxybenzoate CAS:2150-43-8 Purity:>98%
SDP
Catalog No. 501934711
Encompass_Preferred
Medchemexpress, HY-Z0548 50mg Methyl 3,4-dihydroxybenzoate CAS:2150-43-8 Methyl 3,4-dihydroxybenzoate (Protocatechuic acid methyl ester; Methyl protocatechuate) is a major metabolite of antioxidant polyphenols found in green tea. Antioxidant and anti-inflammatory effect [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4711 Supplier Medchemexpress LLC Supplier No. HYZ054850MG
Medchemexpress, HY-Z0548 50mg Methyl 3,4-dihydroxybenzoate CAS:2150-43-8 Methyl 3,4-dihydroxybenzoate (Protocatechuic acid methyl ester; Methyl protocatechuate) is a major metabolite of antioxidant polyphenols found in green tea. Antioxidant and anti-inflammatory effect [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.