Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 3,4-O-Isopropylidene-L-Threonate

Catalog No. 3921705G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-217-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-217-05G Supplier Sigma Aldrich Supplier No. 3921705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to prepare a dihydrofuran substrate in the total synthesis of (-)-echinosporin.

Specifications

CAS 3449-26-1
Density 0.985 g/mL at 25°C
Boiling Point 96°C to 100°C (0.1 mmHg, lit.)
Molecular Formula C16H23NSi2
Refractive Index n20/D 1.446
Linear Formula [C6H5Si(CH3)2]2NH
Quantity 5 g
MDL Number MFCD00042916
Synonym (alphaR,4S)-2,2-Dimethyl-alpha-hydroxy-1,3-dioxolane-4-acetic acid methyl ester; 3,4-O-Isopropylidene-L-threonic acid methyl ester
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.