Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,5-dichlorobenzoate, 98%, Thermo Scientific™

Catalog No. AAB2531106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2531106 5 g
AAB2531114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2531106 Supplier Thermo Scientific Chemicals Supplier No. B2531106

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-67-1
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.03
MDL Number MFCD00032710
InChI Key BTEVDFJXGLQUDS-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzoic acid methyl ester, benzoic acid, 3,5-dichloro-, methyl ester, unii-4gim06ee3p, 4gim06ee3p, methyl-3,5-dichlorobenzoate, maybridge1_004559, acmc-1cd9x, dsstox_cid_22146, dsstox_rid_79937, dsstox_gsid_42146
PubChem CID 76192
IUPAC Name methyl 3,5-dichlorobenzoate
SMILES COC(=O)C1=CC(Cl)=CC(Cl)=C1

Specifications

Melting Point 58°C to 60°C
Quantity 5 g
Formula Weight 205.04
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dichlorobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.