Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-amino-2-hydroxybenzoate, 97%, Thermo Scientific™

Catalog No. MAY00290DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAY00290DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00290DA Supplier Thermo Scientific Chemicals Supplier No. MAY00290DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35748-34-6
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD09842614
InChI Key OMWQHVRUXLRZRC-UHFFFAOYSA-N
Synonym 3-amino-2-hydroxybenzoic acid methyl ester, benzoic acid, 3-amino-2-hydroxy-, methyl ester, methyl 3-aminosalicylate, methyl3-amino-2-hydroxybenzoate, 2-amino-6-methoxycarbonyl phenol, 2-hydroxy-3-methoxycarbonyl aniline, methyl-m-aminosalicylat, acmc-209wuz, methyl-3-aminosalicylate, aminohydroxy-benzoic methyl ester
PubChem CID 12782444
IUPAC Name methyl 3-amino-2-hydroxybenzoate
SMILES COC(=O)C1=C(O)C(N)=CC=C1

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 167.16
Chemical Name or Material Methyl 3-amino-2-hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.