Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Amino-4-chlorobenzoate 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
40872-87-5
C8H8ClNO2
185.607
LOCJPOYKBUUVKU-UHFFFAOYSA-N
3-Amino-4-chlorobenzoic Acid Methyl Ester
593055
methyl 3-amino-4-chlorobenzoate
COC(=O)C1=CC(=C(C=C1)Cl)N
40872-87-5
C8H8ClNO2
185.607
LOCJPOYKBUUVKU-UHFFFAOYSA-N
3-Amino-4-chlorobenzoic Acid Methyl Ester
593055
methyl 3-amino-4-chlorobenzoate
COC(=O)C1=CC(=C(C=C1)Cl)N
Color Yellow
Melting Point 83°C
Quantity 25g
Formula Weight 185.61
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Methyl 3-Amino-4-chlorobenzoate

EINECSNumber : (3)-1481

TSCA : No