Learn More
Medchemexpress LLC Methyl 3-aminopropanoate (hydrochloride) | 3196-73-4 | 139.58 | 10 G
SDP
Catalog No. 5000445814
Encompass_Preferred
Methyl 3-aminopropanoate (hydrochloride), also known as β-Alanine methyl ester hydrochloride, is a compound prepared by the esterification of Β-Alanine (I). It is a white to off-white solid used in the synthesis of bidentate pyridine-acid ligands and for proteomics research.
  • Prepared by esterification of Β-Alanine (I)
  • Used in synthesis of bidentate pyridine-acid ligands
  • Utilized for proteomics research
Catalog No. 50-004-45814 Supplier Medchemexpress LLC Supplier No. HYW00764810G
Methyl 3-aminopropanoate (hydrochloride), also known as β-Alanine methyl ester hydrochloride, is a compound prepared by the esterification of Β-Alanine (I). It is a white to off-white solid used in the synthesis of bidentate pyridine-acid ligands and for proteomics research.
  • Prepared by esterification of Β-Alanine (I)
  • Used in synthesis of bidentate pyridine-acid ligands
  • Utilized for proteomics research