Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Bromo-2-methylbenzoate 98.0+%, TCI America™
SDP

Catalog No. M25435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25435G 5 g
M254325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25435G Supplier TCI America Supplier No. M25435G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99548-54-6
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD09031772
InChI Key MKQQTCSBXHAYQL-UHFFFAOYSA-N
Synonym 3-Bromo-2-methylbenzoic Acid Methyl Ester, Methyl 3-Bromo-o-toluate, 3-Bromo-o-toluic Acid Methyl Ester
PubChem CID 12929135
IUPAC Name methyl 3-bromo-2-methylbenzoate
SMILES CC1=C(C=CC=C1Br)C(=O)OC

Specifications

Melting Point 32°C
Color White-Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 3-Bromo-2-methylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.