Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Bromo-4-fluorobenzoate 98.0+%, TCI America™
SDP

Catalog No. M30181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30181G 1 g
M30185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30181G Supplier TCI America Supplier No. M30181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82702-31-6
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.04
MDL Number MFCD04972915
InChI Key JVORYGNKYAXATM-UHFFFAOYSA-N
Synonym 3-Bromo-4-fluorobenzoic Acid Methyl Ester
PubChem CID 7018022
IUPAC Name methyl 3-bromo-4-fluorobenzoate
SMILES COC(=O)C1=CC=C(F)C(Br)=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 233.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 3-Bromo-4-fluorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.