Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Bromopyruvate 97.0+%, TCI America™
SDP

Catalog No. M22235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22235G Supplier TCI America Supplier No. M22235G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7425-63-0
Molecular Formula C4H5BrO3
Molecular Weight (g/mol) 180.99
MDL Number MFCD01862963
InChI Key MQONVZMIFQQQHA-UHFFFAOYSA-N
Synonym 3-Bromo-2-oxopropionic Acid Methyl Ester, 3-Bromopyruvic Acid Methyl Ester, Methyl 3-Bromo-2-oxopropionate
PubChem CID 352524
IUPAC Name methyl 3-bromo-2-oxopropanoate
SMILES COC(=O)C(=O)CBr

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 84°C
Quantity 5 g
Formula Weight 180.99
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Bromopyruvate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.