Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 3-butenoate

Catalog No. 3842671G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-426-71G 1 g
38-426-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-426-71G Supplier Sigma Aldrich Supplier No. 3842671G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl 3-butenoate is an olefin ester
  • It is reported to undergo iron carbonyl-promoted isomerization to afford α, α-unsaturated esters
  • It is one of the reaction products formed during flash vacuum thermolysis of ()-cocaine
  • The h2 and ch4 chemical ionization mass spectra of methyl 3-butenoate has been reported.methyl 3-butenoate may be employed for the synthesis of dipeptide olefin isosteres using intermolecular olefin cross-metathesis

Specifications

CAS 3724-55-8
Density 0.939 g/mL (at 25°C (literature))
Boiling Point 112°C (lit.)
Molecular Formula C5H8O2
Refractive Index n20/D 1.409 (literature)
Linear Formula CH2=CHCH2COOCH3
MDL Number MFCD00082587
Quantity 1 g
Synonym Methyl vinylacetate
Molecular Weight (g/mol) 100.12
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.