Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 3-carbamoylbicyclo[1.1.1]pentane-1-carboxylate | 156329-77-0 | MFCD20701193 | >97.0% | 169.18 g·mol⁻1 | C8H11NO3 | 250 MG
SDP

Catalog No. 5000303469
Encompass_Preferred

Methyl 3-carbamoylbicyclo[1.1.1]pentane-1-carboxylate is a small-molecule drug intermediate used in medicinal chemistry for the synthesis of bioactive compounds. It is supplied in milligram-scale quantities for research applications and is characterized by the chemical formula C8H11NO3 and a molecular weight of 169.18 g·mol⁻1.

  • Used as a drug intermediate for synthesis of active compounds.
  • Available in small packaging sizes suitable for laboratory research.
  • Typical supplier-reported purity ranges from 95% to ≥97%.
  • Characterized by formula C8H11NO3 and molecular weight 169.18 g·mol⁻1.
  • Intended for research use; follow standard storage and handling procedures.

Catalog No. 50-003-03469 Supplier Medchemexpress LLC Supplier No. 250MGHYW032995
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 3-carbamoylbicyclo[1.1.1]pentane-1-carboxylate is a small-molecule drug intermediate used in medicinal chemistry for the synthesis of bioactive compounds. It is supplied in milligram-scale quantities for research applications and is characterized by the chemical formula C8H11NO3 and a molecular weight of 169.18 g·mol⁻1.

  • Used as a drug intermediate for synthesis of active compounds.
  • Available in small packaging sizes suitable for laboratory research.
  • Typical supplier-reported purity ranges from 95% to ≥97%.
  • Characterized by formula C8H11NO3 and molecular weight 169.18 g·mol⁻1.
  • Intended for research use; follow standard storage and handling procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.