Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-chloro-4-methylbenzoate, 95%

Catalog No. AAH2593314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2593314 25 g
AAH2593306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2593314 Supplier Thermo Scientific Chemicals Supplier No. H2593314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 56525-63-4 | C9H9ClO2 | 184.619 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56525-63-4
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00205141
InChI Key KTFQDZCNPGFKAH-UHFFFAOYSA-N
Synonym methyl3-chloro-4-methylbenzoate, methyl-3-chloro-4-methylbenzoate, methyl 3-chloro-p-toluate, 2-chloro-4-methoxycarbonyl toluene, 3-chloro-4-methylbenzoic acid methyl ester, benzoic acid, 3-chloro-4-methyl-, methyl ester, pubchem10935, acmc-209lsv, ksc495i9j, methyl-3-chloro-4-metylbenzoate
PubChem CID 2801405
IUPAC Name methyl 3-chloro-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)Cl

Specifications

Melting Point 27°C to 29°C
Boiling Point 119°C to 120°C (10 mmHg)
Quantity 25 g
Formula Weight 184.62
Percent Purity 95%
Chemical Name or Material Methyl 3-chloro-4-methylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.