Learn More
Accela Chembio Inc Methyl 3-(chlorosulfonyl)benzoate | 25g | 63555-50-0 | MFCD06408800 | 95+% | Shelf Life: 1440 Days | Moisture Sensitive/light Sensitive/+4
Supplier:  Accela Chembio Inc SY00295125G
Encompass_Preferred
Methyl 3-(chlorosulfonyl)benzoate | 25g | 63555-50-0 | MFCD06408800 | 95+% | Shelf Life: 1440 Days | Moisture Sensitive/light Sensitive/+4
Catalog No. 50-353-578