Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-cyclopentenecarboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AC457400050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457400050 5 g
AC457400010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457400050 Supplier Thermo Scientific Chemicals Supplier No. 457400050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 58101-60-3 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58101-60-3
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD04038661
InChI Key CEOILRYKIJRPBZ-UHFFFAOYSA-N
Synonym methyl 3-cyclopentenecarboxylate, methyl cyclopent-3-enecarboxylate, methyl-3-cyclopentene-1-carboxylate, methyl3-cyclopentenecarboxylate, 3-cyclopentene-1-carboxylic acid methyl ester, 3-cyclopentene-1-carboxylic acid, methyl ester, acmc-209m2j, methyl cyclopent-3-ene carboxylate, 4-methoxycarbonyl cyclopent-1-ene, methyl 3-cyclopentene-1-carboxylate
PubChem CID 1514118
IUPAC Name methyl cyclopent-3-ene-1-carboxylate
SMILES COC(=O)C1CC=CC1

Specifications

Boiling Point 149.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material Methyl 3-cyclopentenecarboxylate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.