Learn More
Accela Chembio Inc Methyl 3-formylindole-6-carboxylate | 1g | 133831-28-4 | MFCD00216479 | 97+% | Shelf Life: 2160 Days | Light Sensitive/nitrogen Or Argon/+4
Supplier:  Accela Chembio Inc SY0036381G
Encompass_Preferred
Methyl 3-formylindole-6-carboxylate | 1g | 133831-28-4 | MFCD00216479 | 97+% | Shelf Life: 2160 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-250-202