Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 3-hydroxy-2-methylenebutyrate

Catalog No. 4046675ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-466-75ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-466-75ML Supplier Sigma Aldrich Supplier No. 4046675ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl 3-hydroxy-2-methylenebutyrate

Specifications

CAS 18020-65-0
Density 1.071 g/mL (at 25°C)
Boiling Point 91°C (20 mmHg)
Molecular Formula C6H10O3
Refractive Index n20/D 1.452 (literature)
Linear Formula CH3CH(OH)C(=CH2)CO2CH3
Quantity 5 mL
MDL Number MFCD00192302
Synonym Methyl 2-(1-hydroxyethyl)acrylate
Molecular Weight (g/mol) 130.14
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.