Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Hydroxy-2-pyridinecarboxylate 98.0+%, TCI America™
SDP

Catalog No. M29735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29735G 5 g
M29731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29735G Supplier TCI America Supplier No. M29735G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62733-99-7
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00661311
InChI Key MHKKUZDJUGIOBC-UHFFFAOYSA-N
Synonym 3-Hydroxy-2-pyridinecarboxylic Acid Methyl Ester, Methyl 3-Hydroxypicolinate, 3-Hydroxypicolinic Acid Methyl Ester
PubChem CID 819341
IUPAC Name methyl 3-hydroxypyridine-2-carboxylate
SMILES COC(=O)C1=C(O)C=CC=N1

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 5 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Hydroxy-2-pyridinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.