Learn More
Accela Chembio Inc Methyl 3-hydroxy-4-methoxybenzoate | 5g | 6702-50-7 | MFCD01321262 | 98% | Shelf Life: 1620 Days | Light Sensitive
Catalog No. 50250766
Encompass_Preferred
Methyl 3-hydroxy-4-methoxybenzoate | 5g | 6702-50-7 | MFCD01321262 | 98% | Shelf Life: 1620 Days | Light Sensitive
Catalog No. 50-250-766 Supplier Accela Chembio Inc Supplier No. SY0053655G
Methyl 3-hydroxy-4-methoxybenzoate | 5g | 6702-50-7 | MFCD01321262 | 98% | Shelf Life: 1620 Days | Light Sensitive